Anti-CD44 Mouse Monoclonal Antibody [clone: B-F24]

Supplier: Cell Sciences
CDM041
102995-502EA 443.08 USD
102995-502
Anti-CD44 Mouse Monoclonal Antibody [clone: B-F24]
Antibodies
Flow Cytometry: Use 10 µl to label 1x10E6 cells or 100 µl of whole blood.

Type: Primary
Antigen: CD44
Clonality: Monoclonal
Clone: B-F24
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR