Anti-CD117 (c-kit) Rat Monoclonal Antibody (Brilliant Violet® 650) [clone: ACK2]

Supplier: BioLegend
135125 135125
10756-414EA 333.55 USD
CA10756-414 10756-414
Anti-CD117 (c-kit) Rat Monoclonal Antibody (Brilliant Violet® 650) [clone: ACK2]
Antibodies
Brilliant Violet 650™ anti-mouse CD117 (c-kit) [ACK2]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC; Size: 50 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.5 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD117 (c-kit)
Clonality: Monoclonal
Clone: ACK2
Conjugation: Brilliant Violet® 650
Epitope:
Host: Rat
Isotype: Rat IgG2b, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR