Anti-CD44 Mouse Monoclonal Antibody [clone: B-F24]

Supplier: Cell Sciences
CDM039
102995-498EA 553.85 USD
102995-498
Anti-CD44 Mouse Monoclonal Antibody [clone: B-F24]
Antibodies
Triggers T lymphocyte aggregation. Enhances hyaluronan binding to CD44.

Type: Primary
Antigen: CD44
Clonality: Monoclonal
Clone: B-F24
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR