Anti-IL13RA1 Mouse Monoclonal Antibody [clone: B-K19]

Supplier: Cell Sciences
CDM022
102995-464EA 540 USD
102995-464
Anti-IL13RA1 Mouse Monoclonal Antibody [clone: B-K19]
Antibodies
Inhibition of IL-13 activation on monocytes (decreasing of IL-6 production).

Type: Primary
Antigen: IL13RA1
Clonality: Monoclonal
Clone: B-K19
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR