Anti-CD117 Mouse Monoclonal Antibody (Brilliant Violet® 510) [clone: 104D2]

Supplier: BioLegend
313219 313220 313219 313220
10758-114EA 487.84 USD
CA10758-114 10758-114 10758-112 CA10758-112
Anti-CD117 Mouse Monoclonal Antibody (Brilliant Violet® 510) [clone: 104D2]
Antibodies
Brilliant Violet 510™ anti-human CD117 (c-kit) [104D2]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-Reactivity: Cynomolgus, Cattle (Bovine, Cow); Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD117
Clonality: Monoclonal
Clone: 104D2
Conjugation: Brilliant Violet® 510
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human;Cross-Reactivity: Cynomolgus;Cattle (Bovine;Cow)
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR