Anti-TNF Mouse Monoclonal Antibody [clone: B-F7]

Supplier: Cell Sciences
CDM412
102995-434EA 1267.42 USD
102995-434
Anti-TNF Mouse Monoclonal Antibody [clone: B-F7]
ELISA: 1 - 10 µg/ml ELISpot: 5 - 10 µg/ml ELISA Capture Antibody. This Antibody can be used as Capture in a human TNF-a sandwich Immunoassay to detect human TNF-a in combination with biotinylated human TNF-a Detection Antibody (Cat n° CDM436). The suggested coating concentration range below should be optimized by each laboratory for each application.

Type: Primary
Antigen: TNF
Clonality: Monoclonal
Clone: B-F7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR