Anti-IL6 Mouse Monoclonal Antibody (Biotin) [clone: B-E4]

Supplier: Cell Sciences
CDM426
102995-436EA 886.15 USD
102995-436
Anti-IL6 Mouse Monoclonal Antibody (Biotin) [clone: B-E4]
Antibodies
Detection Antibody, ELISA, ELISPOT

Type: Primary
Antigen: IL6
Clonality: Monoclonal
Clone: B-E4
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR