Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CD117 (c-kit) Rat Monoclonal Antibody (Brilliant Violet® 605) [clone: ACK2]

Supplier: Biolegend

135122 135121 135122 135121
10756-408 CA10756-406 CA10756-408 10756-406
Anti-CD117 (c-kit) Rat Monoclonal Antibody (Brilliant Violet® 605) [clone: ACK2]
Brilliant Violet 605™ anti-mouse CD117 (c-kit) [ACK2]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC; Size: 500 μl

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining using the μl sizes, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. For flow cytometric staining using the μg size, the suggested use of this reagent is 0.25 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD117 (c-kit)
Clonality: Monoclonal
Clone: ACK2
Conjugation: Brilliant Violet® 605
Epitope:
Host: Rat
Isotype: Rat IgG2b, κ
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty