Anti-SDC1 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: B-A38]

Supplier: Cell Sciences
CDM367
102995-430EA 692.31 USD
102995-430
Anti-SDC1 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: B-A38]
Antibodies
Flow Cytometry: Use 10 µl to label 10E6 cells or 100 µl of whole blood.

Type: Primary
Antigen: SDC1
Clonality: Monoclonal
Clone: B-A38
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR