Anti-CD115 Rat Monoclonal Antibody (Brilliant Violet® 605) [clone: AFS98]

Supplier: BioLegend
135517 135517
10759-306EA 410.11 USD
CA10759-306 10759-306
Anti-CD115 Rat Monoclonal Antibody (Brilliant Violet® 605) [clone: AFS98]
Antibodies
Brilliant Violet 605™ anti-mouse CD115 (CSF-1R) [AFS98]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 50 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.5 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD115
Clonality: Monoclonal
Clone: AFS98
Conjugation: Brilliant Violet® 605
Epitope:
Host: Rat
Isotype: Rat IgG2a, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR