Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CD107b (LAMP-2) Mouse Monoclonal Antibody [clone: H4B4]

Supplier: Biolegend

354302 354302 354301 354301
354302-BL CA354302-BL 354301-BL CA354301-BL
Anti-CD107b (LAMP-2) Mouse Monoclonal Antibody [clone: H4B4]
Purified anti-human CD107b (LAMP-2) [H4B4]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: ICFC, IHC-P, IF; Size: 100 μg

Each lot of this antibody is quality control tested by intracellular immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.5 μg per million cells in 100 μl volume. For immunohistochemical staining on formalin-fixed paraffin-embedded tissue sections, the suggested use of this reagent is 5.0 - 10 μg per ml. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD107b (LAMP-2)
Clonality: Monoclonal
Clone: H4B4
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty