Anti-CD107a (LAMP-1) Rat Monoclonal Antibody (Alexa Fluor® 594) [clone: 1D4B]

Supplier: BioLegend
121622 121622
10757-006EA 273.08 USD
CA10757-006 10757-006
Anti-CD107a (LAMP-1) Rat Monoclonal Antibody (Alexa Fluor® 594) [clone: 1D4B]
Antibodies
Alexa Fluor® 594 anti-mouse CD107a (LAMP-1) [1D4B]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: IF; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescence staining. For immunofluorescence microscopy, a concentration range of 2.5-10 μg/ml is recommended. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD107a (LAMP-1)
Clonality: Monoclonal
Clone: 1D4B
Conjugation: Alexa Fluor® 594
Epitope:
Host: Rat
Isotype: Rat IgG2a, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR