Anti-CD107a Mouse Monoclonal Antibody (Brilliant Violet® 605) [clone: H4A3]

Supplier: BioLegend
328634 328633 328633
10019-308EA 561.43 USD
10019-308 10758-380 CA10758-380
Anti-CD107a Mouse Monoclonal Antibody (Brilliant Violet® 605) [clone: H4A3]
Antibodies
Brilliant Violet 605™ anti-human CD107a (LAMP-1) [H4A3]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Chimpanzee, Cynomolgus, Pigtailed Macaque, Rhesus; Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD107a
Clonality: Monoclonal
Clone: H4A3
Conjugation: Brilliant Violet® 605
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human;African Green;Baboon;Chimpanzee;Cynomolgus;Pigtailed Macaque;Rhesus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR