Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CD104 Rat Monoclonal Antibody (Alexa Fluor® 647) [clone: 346-11A]

Supplier: Biolegend

123607 123608 123607 123608
10759-260EA 147.52 USD InStock
10759-260 10759-262 CA10759-260 CA10759-262
Anti-CD104 Rat Monoclonal Antibody (Alexa Fluor® 647) [clone: 346-11A]
Antibodies
Alexa Fluor® 647 anti-mouse CD104 [346-11A]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.5 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD104
Clonality: Monoclonal
Clone: 346-11A
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rat
Isotype: Rat IgG2a, κ
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty