Anti-CD103 Mouse Monoclonal Antibody (Biotin) [clone: OX-62]

Supplier: BioLegend
205503 205503
205503-BLEA 165.19 USD
CA205503-BL 205503-BL
Anti-CD103 Mouse Monoclonal Antibody (Biotin) [clone: OX-62]
Antibodies
Biotin anti-rat CD103 (αE Integrin) [OX-62]; Isotype: Mouse IgG1; Reactivity: Rat; Apps: FC, ICC, IHC, WB; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 1.0 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD103
Clonality: Monoclonal
Clone: OX-62
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: Mouse IgG1
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR