Anti-CD103 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: Ber-ACT8]

Supplier: BioLegend
350215 350216 350215 350216
10758-950EA 137.15 USD
CA10758-950 10758-950 CA10758-952 10758-952
Anti-CD103 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: Ber-ACT8]
Antibodies
APC anti-human CD103 (Integrin αE) [Ber-ACT8]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-Reactivity: Cynomolgus; Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD103
Clonality: Monoclonal
Clone: Ber-ACT8
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human;Cross-Reactivity: Cynomolgus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR