Anti-CD103 Armenian Hamster Monoclonal Antibody (Brilliant Violet® 510) [clone: 2E7]

Supplier: BioLegend
121423 121423
10756-996EA 333.47 USD
10756-996 CA10756-996
Anti-CD103 Armenian Hamster Monoclonal Antibody (Brilliant Violet® 510) [clone: 2E7]
Antibodies
Brilliant Violet 510™ anti-mouse CD103 [2E7]; Isotype: Armenian Hamster IgG; Reactivity: Mouse; Apps: FC; Size: 50 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.5 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD103
Clonality: Monoclonal
Clone: 2E7
Conjugation: Brilliant Violet® 510
Epitope:
Host: Armenian Hamster
Isotype: Armenian Hamster IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR