Anti-CD100 Rat Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: BMA-12]

147607 147608 147608 147607
10756-600EA 212.67 USD
CA10756-600 CA10759-214 10756-600 10759-214
Anti-CD100 Rat Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: BMA-12]
FITC anti-mouse CD100 (SEMA4D) [BMA-12]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 1.0 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD100
Clonality: Monoclonal
Clone: BMA-12
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rat
Isotype: Rat IgG2a, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR