Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CD100 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: BMA-12]

Supplier: Biolegend

147605 147606 147605 147606
10759-210EA 147.52 USD InStock
10759-210 CA10759-212 CA10759-210 10759-212
Anti-CD100 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: BMA-12]
Antibodies
APC anti-mouse CD100 (SEMA4D) [BMA-12]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 1.0 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD100
Clonality: Monoclonal
Clone: BMA-12
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rat
Isotype: Rat IgG2a, κ
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty