Anti-CD117 Mouse Monoclonal Antibody [clone: B-K15]

Supplier: Cell Sciences
CDM205
102995-412EA 443.08 USD
102995-412
Anti-CD117 Mouse Monoclonal Antibody [clone: B-K15]
Antibodies
Flow Cytometry: Use 10 µl to label 1x10E6 cells or 100 µl of whole blood.

Type: Primary
Antigen: CD117
Clonality: Monoclonal
Clone: B-K15
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR