Anti-CCX-CKR Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 13E11]

Supplier: BioLegend
362103 362104 362103 362104
10760-770EA 361.57 USD
10760-770 CA10760-768 10760-768 CA10760-770
Anti-CCX-CKR Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 13E11]
Antibodies
PE anti-human CCX-CKR (CCRL1) [13E11]; Isotype: Mouse IgG2b, κ; Reactivity: Human; Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CCX-CKR
Clonality: Monoclonal
Clone: 13E11
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: Mouse IgG2b, κ
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR