Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-TPTE2 Mouse Monoclonal Antibody [clone: 978CT6.1.4]

Supplier: Abgent

AM2224A
10217-718
Anti-TPTE2 Mouse Monoclonal Antibody [clone: 978CT6.1.4]
Western Blot: 1:5000

Type: Primary
Antigen: TPTE2
Clonality: Monoclonal
Clone: 978CT6.1.4
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty