Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CCL28 Rat Monoclonal Antibody [clone: L019H7]

Supplier: Biolegend

525802 525802
10761-794EA 333.28 USD InStock
10761-794 CA10761-794
Anti-CCL28 Rat Monoclonal Antibody [clone: L019H7]
Antibodies
Purified anti-mouse CCL28 (MEC) [L019H7]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: ELISA Capture; Size: 500 μg

Each lot of this antibody is quality control tested by ELISA assay. For ELISA capture applications, a concentration range of 1-4 μg/mL is recommended. To obtain a linear standard curve, serial dilutions of mouse CCL28 recombinant protein ranging from 1000 to 15.6 pg/mL are recommended for each ELISA plate. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CCL28
Clonality: Monoclonal
Clone: L019H7
Conjugation:
Epitope:
Host: Rat
Isotype: Rat IgG2a, κ
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty