Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CCL28 Mouse Monoclonal Antibody (Biotin) [clone: J139G11]

Supplier: Biolegend

525703 525703
10761-792EA 163.91 USD InStock
10761-792 CA10761-792
Anti-CCL28 Mouse Monoclonal Antibody (Biotin) [clone: J139G11]
Antibodies
Biotin anti-human CCL28 (MEC) [J139G11]; Isotype: Mouse IgG2b, κ; Reactivity: Human; Apps: ELISA Detection; Size: 50 μg

Each lot of this antibody is quality control tested by ELISA assay. For ELISA detection applications, a concentration range of 0.1-0.5 μg/mL is recommended. To obtain a linear standard curve, serial dilutions of human CCL28 recombinant protein ranging from 1000 to 15.6 pg/mL are recommended for each ELISA plate. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CCL28
Clonality: Monoclonal
Clone: J139G11
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: Mouse IgG2b, κ
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty