Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CCL28 Goat Polyclonal Antibody (Biotin) [clone: Poly5242]

Supplier: Biolegend

524203 524203
10762-154EA 278.65 USD InStock
10762-154 CA10762-154
Anti-CCL28 Goat Polyclonal Antibody (Biotin) [clone: Poly5242]
Antibodies
Biotin anti-mouse CCL28 (MEC) [Poly5242]; Isotype: Goat Polyclonal Ig; Reactivity: Mouse; Apps: ELISA Detection; Size: 50 μg

Each lot of this antibody is quality control tested by ELISA assay. For ELISA detection applications, a concentration range of 0.1-0.2 μg/mL is recommended. To obtain a linear standard curve, serial dilutions of mouse CCL28 recombinant protein ranging from 1000 to 15.6 pg/mL are recommended for each ELISA plate. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CCL28
Clonality: Polyclonal
Clone: Poly5242
Conjugation: Biotin
Epitope:
Host: Goat
Isotype: Goat Polyclonal Ig
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty