Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CCL17 Goat Polyclonal Antibody (Biotin) [clone: Poly5230]

Supplier: Biolegend

523003 523003
10762-004 CA10762-004
Anti-CCL17 Goat Polyclonal Antibody (Biotin) [clone: Poly5230]
Biotin anti-human CCL17 (TARC) [Poly5230]; Isotype: Goat Polyclonal Ig; Reactivity: Human; Apps: ELISA Detection; Size: 50 μg

Each lot of this antibody is quality control tested by ELISA assay. For use as an ELISA detection antibody, a concentration range of 0.25-0.5 μg/ml is recommended. To obtain a linear standard curve, serial dilutions of CCL17 (TARC) recombinant protein ranging from 500 to 7.8 pg/ml are recommended for each ELISA plate. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CCL17
Clonality: Polyclonal
Clone: Poly5230
Conjugation: Biotin
Epitope:
Host: Goat
Isotype: Goat Polyclonal Ig
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty