Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CCL11 Rat Monoclonal Antibody [clone: L402H11]

Supplier: Biolegend

660402 660402
10763-352EA 265.23 USD InStock
10763-352 CA10763-352
Anti-CCL11 Rat Monoclonal Antibody [clone: L402H11]
Antibodies
Purified anti-CCL11 (Eotaxin) [L402H11]; Isotype: Rat IgG2a; Reactivity: Mouse; Apps: WB; Size: 100 μg

Each lot of this antibody is quality control tested by Western blotting. For Western blotting, the suggested use of this reagent is 0.5 - 2.0 μg per ml. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CCL11
Clonality: Monoclonal
Clone: L402H11
Conjugation:
Epitope:
Host: Rat
Isotype: Rat IgG2a
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty