Anti-CCL11 Rat Monoclonal Antibody [clone: 16D10A40]

Supplier: BioLegend
656701 656701
10763-648EA 316.31 USD
10763-648 CA10763-648
Anti-CCL11 Rat Monoclonal Antibody [clone: 16D10A40]
Antibodies
Purified anti-CCL11 (Eotaxin) [16D10A40]; Isotype: Rat IgG2a, κ; Reactivity: Human; Apps: WB; Size: 500 μg

Each lot of this antibody is quality control tested by Western blotting. For Western blotting, the suggested use of this reagent is 0.5 - 2.0 μg per ml. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CCL11
Clonality: Monoclonal
Clone: 16D10A40
Conjugation:
Epitope:
Host: Rat
Isotype: Rat IgG2a, κ
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR