Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Bin1 Mouse Monoclonal Antibody [clone: 99D]

Supplier: Biolegend

655602 655602
CA10763-534 10763-534
Anti-Bin1 Mouse Monoclonal Antibody [clone: 99D]
Antibodies
Purified anti-Bin1 [99D]; Isotype: Mouse IgG2b; Reactivity: Human; Apps: WB, IP, FC, ELISA; Size: 100 μg

Each lot of this antibody is quality control tested by Western blotting. For Western blotting, the suggested use of this reagent is 0.5 - 4.0 μg per ml. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: Bin1
Clonality: Monoclonal
Clone: 99D
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse IgG2b
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty