Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Bcl-6 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 7D1]

Supplier: Biolegend

358506 358505 358506 358505
CA10760-442 CA10760-440 10760-442 10760-440
Anti-Bcl-6 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 7D1]
APC anti-human/mouse Bcl-6 [7D1]; Isotype: Rat IgG2a, κ; Reactivity: Human, Mouse; Apps: ICFC; Size: 100 tests

Each lot of this antibody is quality control tested by intracellular immunofluorescent staining using our nuclear factor staining protocol. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: Bcl-6
Clonality: Monoclonal
Clone: 7D1
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rat
Isotype: Rat IgG2a, κ
Reactivity: Human;Mouse

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty