Anti-CLEC9A Rabbit Polyclonal Antibody

Supplier: Abcepta
AP17684C
89517-718EA 636.79 USD
89517-718
Anti-CLEC9A Rabbit Polyclonal Antibody
Antibodies
Western Blot: 1:1000

Type: Primary
Antigen: CLEC9A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Center
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR