Anti-Bax Mouse Monoclonal Antibody (Alexa Fluor® 488) [clone: 2D2]

Supplier: BioLegend
633603 633604 633603 633604
10763-220EA 178.17 USD
CA10763-220 10763-220 10763-222 CA10763-222
Anti-Bax Mouse Monoclonal Antibody (Alexa Fluor® 488) [clone: 2D2]
Antibodies
Alexa Fluor® 488 anti-Bax [2D2]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: ICFC; Size: 100 μg

Each lot of this antibody is quality control tested by intracellular immunofluorescent staining using our nuclear factor staining protocol. For flow cytometric staining, the suggested use of this reagent is 1.0 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: Bax
Clonality: Monoclonal
Clone: 2D2
Conjugation: Alexa Fluor® 488
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR