Anti-B7-H4 Mouse Monoclonal Antibody (PE (Phycoerythrin)/Cy7®) [clone: MIH43]

Supplier: BioLegend
358106 358106 358105 358105
10760-326EA 412.26 USD
CA10760-324 CA10760-326 10760-326 10760-324
Anti-B7-H4 Mouse Monoclonal Antibody (PE (Phycoerythrin)/Cy7®) [clone: MIH43]
Antibodies
PE/Cy7 anti-human B7-H4 [MIH43]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: B7-H4
Clonality: Monoclonal
Clone: MIH43
Conjugation: PE (Phycoerythrin)/Cy7®
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR