Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-ATM Phospho (Ser1981) Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 10H11.E12]

Supplier: Biolegend

651203 651203 651204 651204
10763-406EA 158.44 USD InStock
10763-406 CA10763-406 CA10763-408 10763-408
Anti-ATM Phospho (Ser1981) Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 10H11.E12]
Antibodies
PE anti-ATM Phospho (Ser1981) [10H11.E12]; Isotype: Mouse IgG1, κ; Reactivity: Human, Mouse; Apps: ICFC; Size: 100 μg

Each lot of this antibody is quality control tested by intracellular immunofluorescent staining using our nuclear factor staining protocol. For flow cytometric staining, the suggested use of this reagent is 0.125 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: ATM Phospho (Ser1981)
Clonality: Monoclonal
Clone: 10H11.E12
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human;Mouse

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty