Anti-APRIL Mouse Monoclonal Antibody [clone: 33H5]

Supplier: BioLegend
524801 524801 524802 524802
10761-770EA 388.83 USD
10761-770 CA10761-768 10761-768 CA10761-770
Anti-APRIL Mouse Monoclonal Antibody [clone: 33H5]
Antibodies
Purified anti-human APRIL (TNFSF13, CD256) [33H5]; Isotype: Mouse IgG2a, κ; Reactivity: Human; Apps: ELISA Capture; Size: 500 μg

<span style="color: #333333; font-family: Arial, Helvetica, sans-serif; font-size: 12.222222328186035px; font-style: normal; font-variant: normal; font-weight: normal; letter-spacing: normal; line-height: 17.77777862548828px; orphans: auto; text-align: -webkit-left; text-indent: 0px; text-transform: none; white-space: normal; widows: auto; word-spacing: 0px; -webkit-text-stroke-width: 0px; background-color: #ffffff; display: inline !important; float: none;">Each lot of this antibody is quality control tested by<span class="Apple-converted-space"> </span></span><a style="font-family: Arial, Helvetica, sans-serif; font-size: 12.222222328186035px; color: #4c0073; text-decoration: underline; line-height: 17.77777862548828px; font-style: normal; font-variant: normal; font-weight: normal; letter-spacing: normal; orphans: auto; text-align: -webkit-left; text-indent: 0px; text-transform: none; white-space: normal; widows: auto; word-spacing: 0px; -webkit-text-stroke-width: 0px; background-color: #ffffff;" title="View Protocol" href="http://www.biolegend.com/media_assets/support_protocol/BioLegend_Sandwich_ELISA_protocol.pdf" target="_blank">ELISA assay</a><span style="color: #333333; font-family: Arial, Helvetica, sans-serif; font-size: 12.222222328186035px; font-style: normal; font-variant: normal; font-weight: normal; letter-spacing: normal; line-height: 17.77777862548828px; orphans: auto; text-align: -webkit-left; text-indent: 0px; text-transform: none; white-space: normal; widows: auto; word-spacing: 0px; -webkit-text-stroke-width: 0px; background-color: #ffffff; display: inline !important; float: none;">. </span>For sandwich ELISA capture applications, a concentration range of 2-8 μg/ml is recommended when used in conjunction with biotinylated 53E11 (Cat. No. 524904) as the detection antibody, and recombinant human APRIL (TNFSF13) (ELISA Std.) (Cat. No. 591309) as the protein standard. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: APRIL
Clonality: Monoclonal
Clone: 33H5
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse IgG2a, κ
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR