Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Annexin V N/A N/A Antibody (APC (Allophycocyanin))

Supplier: Biolegend

640919 640920 640920 640919
10763-242 10763-244 CA10763-244 CA10763-242
Anti-Annexin V N/A N/A Antibody (APC (Allophycocyanin))
APC Annexin V Reactivity: All mammalian species; Apps: FC; Size: 100 tests

Each lot of this product is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per 100,000 - million cells in a 100 μl volume of Annexin V Binding Buffer (Cat No. 422201). It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: Annexin V
Clonality: N/A
Clone:
Conjugation: APC (Allophycocyanin)
Epitope:
Host: N/A
Isotype:
Reactivity: All mammalian species

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty