Anti-AMPKa Rabbit Polyclonal Antibody [clone: Poly6498]

Supplier: BioLegend
649802 649802 649801 649801
649802-BLEA 412.82 USD
CA649802-BL CA649801-BL 649802-BL 649801-BL
Anti-AMPKa Rabbit Polyclonal Antibody [clone: Poly6498]
Purified anti-AMPKα [Poly6498]; Isotype: Rabbit Polyclonal IgG; Reactivity: Human; Apps: WB; Size: 200 μl

Each lot of this antibody is quality control tested by <a title="View Protocol" href="http://www.biolegend.com/media_assets/support_protocol/BioLegend_Western_blotting_protocol.pdf" target="_blank">Western blotting</a>. Western blotting, suggested working dilution(s): Use 10 μl per 5 ml antibody dilution buffer for each mini-gel. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: AMPKa
Clonality: Polyclonal
Clone: Poly6498
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit Polyclonal IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR