Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Aiolos Mouse Monoclonal Antibody [clone: 8B2]

Supplier: Biolegend

653201 653201 653202 653202
10763-374EA 131.13 USD InStock
10763-374 CA10763-374 CA10763-376 10763-376
Anti-Aiolos Mouse Monoclonal Antibody [clone: 8B2]
Antibodies
Purified anti-mouse Aiolos [8B2]; Isotype: Mouse IgG1, κ; Reactivity: Mouse; Apps: WB, ICFC, IP; Size: 100 μg

Each lot of this antibody is quality control tested by Western blotting. Western blotting, suggested working dilution(s): Use 0.5 μg/ml antibody dilution buffer for each mini-gel. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: Aiolos
Clonality: Monoclonal
Clone: 8B2
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty