Anti-AIM2 Mouse Monoclonal Antibody [clone: 3B10]

Supplier: BioLegend
652802 652802 652801 652801
10763-318EA 165.19 USD
10763-318 CA10763-318 10763-320 CA10763-320
Anti-AIM2 Mouse Monoclonal Antibody [clone: 3B10]
Antibodies
Purified anti-AIM2 [3B10]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: WB, IF, IP; Size: 100 μg

Each lot of this antibody is quality control tested by Western blotting. For Western blotting, the suggested use of this reagent is 0.5-2.0 μg per ml. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: AIM2
Clonality: Monoclonal
Clone: 3B10
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR