Anti-Acetylated Lysine Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 15G10]

Supplier: BioLegend
623404 623404
10763-204EA 361.57 USD
CA10763-204 10763-204
Anti-Acetylated Lysine Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 15G10]
Antibodies
PE anti-Acetylated Lysine [15G10]; Isotype: Mouse IgG2b, κ; Reactivity: Acetylated lysine residues; Apps: ICFC; Size: 100 tests

Each lot of this antibody is quality control tested by intracellular immunofluorescent staining using our nuclear factor staining protocol. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: Acetylated Lysine
Clonality: Monoclonal
Clone: 15G10
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: Mouse IgG2b, κ
Reactivity: Acetylated lysine residues
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR