Would you like to log in again?
Mechanical Balances, Heavy Duty, Ohaus®
Supplier: OHAUS SCALE CORP
80000000
The Industry Standard for Precision and High Capacity Mechanical Weighing.
  • The oversized stainless steel platform to accommodate large samples or containers
  • Covers protect the self-aligning bearings which require little to no maintenance
  • A lockable Tare feature allows the operator to tare the balance once for multiple weighing procedures for containers up to 2270 g
Durable enough to resist corrosion, this precise balance also weighs with a feather touch, down to 1 g over the whole weighing range - even under the most demanding conditions.
ORDER
Expand All / Collapse All
Weighing Capacity Pan/Platform Size Readability Supplier No. VWR Catalog Number Unit Price Quantity
20000 g (44.09 lbs.) 279 mm 1 g (0.002 lbs.) 80000000 12395-085 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Q and A
Product Questions & Answers
Sort by: Most Helpful | Newest | Oldest
Items Per Page: 5  10  20 
2  3  4  Next
Can't find an answer to your question?  Ask us a question
Ask us a question:
Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question
Q&A Guidelines & Tips
If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.
When writing your question or answer, please follow these guidelines:
  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English
All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s
We reserve the right not to post your question if it contains any of the following
  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty