Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Allure® Guard Cartridges, Restek

Supplier: RESTEK CORP
Allure®
916950222 916950212 916550210 916950210 916550212 915950210
Allure® guard cartridges protect HPLC columns as well as remove particulate contaminants and highly adsorptive compounds from samples, prolonging analytical column life.

  • Available in a number of sizes
  • Removes particulate contaminants
  • Prolonged analytical column life

Effective for prolonging analytical column life.




 

ORDER

Expand All / Collapse All
Column Format Phase I.D. x L Particle Size Supplier No. VWR Catalog Number Unit Price Quantity
Guard cartridge Allure PFP Propyl 2.1 x 10 mm 5 µm 916950212 10840-236 Pack of 1 Retrieving
Guard cartridge Allure Organic Acids 2.1 x 10 mm 5 µm 916550212 10803-450 Pack of 3 Retrieving
Guard cartridge Allure PFP Propyl 2.1 x 20 mm 5 µm 916950222 10840-238 Pack of 1 Retrieving
Guard cartridge Allure AK 4.0 x 10 mm 5 µm 915950210 10840-208 Pack of 1 Retrieving
Guard cartridge Allure Organic Acids 4.0 x 10 mm 5 µm 916550210 10840-224 Pack of 1 Retrieving
Guard cartridge Allure PFP Propyl 4.0 x 10 mm 5 µm 916950210 10840-234 Pack of 1 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty