Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ultra Quat Columns, Restek

Supplier: RESTEK CORP
9181565-700 9181352 9181565
These columns are retentive, high-purity, base-deactivated, reversed-phase packing.

  • Particle Size: 3 µm or 5 µm, spherical
  • Pore Size: 100 Å
  • pH Range: 2.5 to 8
  • Temperature Limit: 80°C

Ideal for the analysis of paraquat and diquat or other quaternary amines.




 

ORDER

Expand All / Collapse All
Description Column Format Phase I.D. x L Packing Material Particle Size Pore Size Supplier No. VWR Catalog Number Unit Price Quantity
Ultra Quat, 3um, 50mm HPLC Columnx2.1mm HPLC Column Analytical Quat 2.1 x 50 mm X 2 Re 3 µm 100 Å 9181352 10839-636 Each Retrieving
Ultra Quat, 5um, 150x4.6mm HPLC Column Analytical Quat 4.6 x 150 mm X 4 Re 5 µm 100 Å 9181565 10839-638 Each Retrieving
Ultra Quat, 5um, 150x4.6mm HPLC Column, with Trident Analytical Quat 4.6 x 150 mm W Re 5 µm 100 Å 9181565-700 10840-880 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty