Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Allure® AK Columns, Restek

Supplier: RESTEK CORP

9159523-700 9159525-700
10838-242EA 831.84 USD InStock
10838-242 10838-244
Allure® AK Columns, Restek
Chromatography Columns
This highly retentive, highly selective phase—unique to Restek—was developed specifically for the analysis of aldehydes and ketones as DNPH derivatives.

  • Particle Size: 5 µm, spherical
  • Pore Size: 60 Å
  • End-Cap: yes
  • pH Range: 2.5 to 8
  • Temperature Limit: 80°C

Allure® AK is a reversed-phase HPLC material that has the unique ability to separate all 13 carbonyl compounds specified in California Air Resources Board (CARB) Method #1004 using a simple acetonitrile/water gradient. Other columns require long analysis times or the use of tetrahydrofuran.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty