Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Pinnacle® DB PAH UHPLC Columns, Restek

Supplier: RESTEK CORP

9470212 9470232 9470252
10837-750EA 745.23 USD InStock
10837-750 10837-752 10837-754
Pinnacle® DB PAH UHPLC Columns, Restek
Chromatography Columns
Pinnacle® DB PAH UHPLC columns are specifically designed to resolve complex mixtures of polycyclic aromatic hydrocarbons (PAHs).

  • Particle Size: 1.9 µm, spherical
  • Pore Size: 140 Å
  • End-Cap: no
  • pH Range: 2.5 to 8
  • Temperature Limit: 80°C

Get complete resolution of all 16 EPA 610 PAHs, plus two other routinely analyzed PAH compounds, in less than five minutes to greatly reduce run times and increase sample throughput.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty