Would you like to log in again?
Human CellExp™ Beta-NGF, Human Recombinant, BioVision
Supplier: BioVision
6443-50 6443-10
10008-590EA 751.9 USD InStock
10008-590 10008-588
Human CellExp™ Beta-NGF, Human Recombinant, BioVision
Proteins and Peptides
>95% Pure HEK 293 cells expressed Human recombinant Beta-NGF
ED₅₀ is typically 1.5 to 8 ng/mL. The specific activity was determined by the dose-dependent stimulation of the proliferation of human TF-1 cells (human erythroleukemic indicator cell line).
NGF-beta has nerve growth stimulating activity and the complex is involved in the regulation of growth and the differentiation of sympathetic and certain sensory neurons. Mutations in this gene have been associated with hereditary sensory and autonomic neuropathy, type 5 (HSAN5), and dysregulation of this gene's expression is associated with allergic rhinitis. The presence of beta NGF in immune cells and endocrine cells as well as in the CNS limbic areas suggests that beta NGF may function as an intracellular messenger to regulate the body’s response to stress. This product is produced in a human cell expression system with serum-free, chemically defined media.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty