Would you like to log in again?
Heaney Needle Holder, OR Grade, Sklar
Supplier: Sklar
20-4682 20-4682C
Sklar's Heaney Needle Holder is a ratcheted, finger ring instrument used for light to medium weight suturing
It is most commonly used in gynecological procedures such as vaginal and abdominal hysterectomy, and dilation and curettage. The slightly curved shanks and tips provide the user a better view when suturing intravaginally, and the cross-serrations provide a secure hold on the needle. This needle holder may also be used as a cardiovascular approximation and closure instrument.
Ordering information: Censitrac instruments are custom barcoded to reflect the facility. This is unique customization. The initial order for the facility can take 14 to 30 days, subsequent orders can take less time.
Caution: Customers must have a Censitrac Site ID to purchase Censitrac instruments.
ORDER
Expand All / Collapse All
Heaney Needle Holder, OR Grade
Description Grade Material Length Sterility Handle Tip Configuration Working Surface Style Curvature Supplier No. VWR Catalog Number Unit Price Quantity
Heaney Needle Holder Premium OR-Grade Stainless Steel 20.9 cm (8.25") Non-Sterile Finger Rings Blunt Cross-Serrated Curved 20-4682 10193-962 Each Retrieving Restricted
restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:
  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address
* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
SPECIFICATIONS
Latex/Latex-free Latex-Free
Disposable/Reusable Reusable
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty