Would you like to log in again?
Human recombinant UBE2E2 (UbcH8), BioVision
Supplier: BioVision
6438-3
10008-576
Human recombinant UBE2E2 (UbcH8), BioVision
>95% Pure Human recombinant UBE2E2 (UbcH8)
Protein concentration of 100 nM - 1 µM is recommended for in vitro conjugation.
Ubiquitin-conjugating enzyme E2 (UBE2E2) is a protein that in humans is encoded by the UBE2E2 gene. Amino acid sequence within the UBC domain of UBE2E2 shares over 90% identity with human UbcH6, mouse UbcM2, and Drosophila UbcD2, whereas the N-terminal region shows little amino acid sequence similarity with known proteins. The UBE2E2 protein forms a thioester bond with ubiquitin in an E1-dependent manner and mediates the transfer of ubiquitin from a ubiquitin-activating enzyme (E1) to a substrate protein or E3 ligase.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty