Would you like to log in again?
Human recombinant UBE2R2 (Ubc3B), BioVision
Supplier: BioVision
6437-3
10008-574EA 324.45 USD InStock
10008-574
Human recombinant UBE2R2 (Ubc3B), BioVision
Proteins and Peptides
>95% Pure Human recombinant UBE2R2 (Ubc3B)
Protein concentration of 100 nM - 1 µM is recommended for in vitro conjugation.
UBE2R2 is a 238 amino acid (27.2 kDa) pre-protein from chromosome 9. After CK2-mediated UBE2R2 phosphorylation, it binds to the F-Box protein beta-TrCP, which is the subunit recognition subunit of an SCF E3 ligase. UBE2R2 may be involved in degradation of beta-catenin.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty