Human Recombinant UBE2L3
Supplier: BioVision
6431-3
10008-046EA 338.58 USD
10008-046
Human Recombinant UBE2L3
>95% Pure Human recombinant UBE2L3 (UbcH7)
Typical enzyme concentration of 100 nM - 1 mM is used for in vitro conjugation, depending on conditions.
UBE2L3 (UbcH7) is an E2 (ubiquitin conjugating enzyme) ligase involved in the conjugation of ubiquitin to target substrates, along with E1 and E3 enzymes. UbcH7 has been shown to work with HECT domain E3’s (E6-AP), in addition to RING containing E3’s. UbcH7 is homologous to the UbcH5 family.